Learn More
Brand™ LDPE Screw Caps for Narrow Mouth Bottles
LDPE Screw Cap that fits GL 25 Thread narrow neck laboratory bottles that are ideal in most research, scientific, and other types of laboratory environments.
Brand:  Brand™ 129204
Additional Details : Poids : 9.00000kg
 View more versions of this product
Product Code. 15241279
  • 13.15€ / Pack of 50
Expédition estimée
01-10-2020
Add to basket
Description
Description
  • LDPE screw cap for narrow mouth bottles
  • Fits GL 25 thread narrow neck laboratory bottles
  • For storage of substances
  • Includes 50 pack of screw caps
Specifications
Specifications
LDPE
Screw Cap
Yes
50Pack
Fits GL 25 Thread narrow neck laboratory bottles