Learn More
BRAND™ LDPE Screw Caps for Wide Mouth Bottles
Catalog Number 15281279
Click to view available options
Thread Size (Metric):
GL 32
GL 40
GL 50
For Use With (Equipment):
Fits GL 40 Thread wide mouth laboratory bottles
Manufactured from Low-Density Polyethylene (LDPE.) BRAND™ LDPE Screw Caps for Wide Mouth Bottles are ridged for easy gripping. Available in a variety of thread sizes. For use with LDPE wide mouth laboratory bottles.
  • Manufactured from Low-Density Polyethylene (LDPE)
  • Ridged for easy gripping
  • Natural color
  • Available in a variety of thread sizes
  • For use with LDPE wide mouth laboratory bottles
Material LDPE
Quantity 25Pack
Type Screw Cap
Thread Size (Metric) GL 50
Certified Yes
For Use With (Equipment) Fits GL 50 Thread wide mouth laboratory bottles