Learn More
Kautex™ LDPE Screw Closure
Product Code. 10353591
Click to view available options
Diameter (Metric):
18 mm
25 mm
28 mm
40 mm
50 mm
65 mm
Unit Size:
Each
Pack of 100
Product Code. 10353591
Brand: Kautex™ 2000070609
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Feature LDPE light weight, tough construction. Kautex™ LDPE Screw Closures are available in a wide variety of sizes for Kautex™ narrow and wide neck bottles.
  • Made from low density polyethylene
  • Translucent
  • Available for a wide variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material LDPE
Thread Size (Metric) 00 mm
Neck Finish 00 mm
For Use With (Equipment) 500/750/1000 mL Wide Mouth Bottle
Diameter (Metric) 50 mm