Learn More
Lead(II) 2,4-pentanedionate, Thermo Scientific Chemicals
Catalog Number p-7080528
Click to view available options
Quantity:
10 g
50 g
15282-88-9
C10H14O4Pb
405.418
MFCD00013499
UNNUWSQNTAFLDC-SYWGCQIGSA-L
lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
53393517
bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C
15282-88-9
C10H14O4Pb
405.418
MFCD00013499
UNNUWSQNTAFLDC-SYWGCQIGSA-L
lead ii acetylacetonate, 2,4-pentanedione lead ii derivative
53393517
bis[[(E)-4-oxopent-2-en-2-yl]oxy]lead
CC(=CC(=O)C)O[Pb]OC(=CC(=O)C)C
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 141°C to 144°C
Odor Odorless
Quantity 10 g
UN Number UN3467
Sensitivity Hygroscopic
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 405.41
Physical Form Crystalline
Chemical Name or Material Lead (II) 2,4-pentanedionate
compliance-icons compliance-icons
Product Identifier
  • Lead(II) 2,4-pentanedionate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360-May damage fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .