missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Acetate Trihydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
Specifications
Melting Point | 75°C |
Color | White |
pH | 5.5 to 6.5 |
Packaging | Plastic powder jar |
Quantity | 250 g |
Formula Weight | 379.33g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Lead(II) acetate trihydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH201-Contains lead. Should not be used on surfaces liable to be chewed or sucked by children.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'