missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) nitrate, Puriss., ≥99%, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
50 kg
Unit Size:
1kg
500g
50kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | Pb(NO3)2 |
| UN Number | 1469 |
| SMILES | [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| IUPAC Name | lead(2+);dinitrate |
| Formula Weight | 331.21g/mol |
| Percent Purity | ≥99% |
| Grade | puriss. |
| Chemical Name or Material | Lead(Ii) Nitrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction