missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Oxide, puriss. p.a., ≥99.0% (KT), Honeywell Fluka™

Catalog Number 15651450
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
1317-36-8
OPb
223.20
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
14827
O=[Pb]
This item is not returnable. View return policy
1317-36-8
OPb
223.20
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
14827
O=[Pb]

Puriss. p.a.,≥99.0% (KT), yellow

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Boiling Point 1470°C
Melting Point 890°C
Color Yellow
Physical Form Solid
Quantity 50 g
Linear Formula PbO
UN Number UN2291
IUPAC Name oxolead
Formula Weight 223.2g/mol
Percent Purity ≥99.0% (KT)
Grade puriss. p.a.
Packaging Poly Bottle
Vapor Pressure 10mmHg (0°C)
Density 9.66g/cu.cm
Chemical Name or Material Lead(Ii) Oxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'