missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(IV) Oxide, ≥97.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Color | Black |
Quantity | 500 g |
Linear Formula | PbO2 |
UN Number | UN1872 |
Formula Weight | 239.2g/mol |
Percent Purity | ≥97.0% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Appearance | Powder |
Chemical Name or Material | Lead(Iv) Oxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'