missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solder, Sn/Pb, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Solder, Sn/Pb, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Iron/Nickel/Mercury/Indium/Bismuth/Cadmium/Arsenic/Silver/Antimony/Copper/Tin/Gold/Zinc/Tellurium |
| CAS | 7440-31-5 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Iron 0.0016 %/Nickel 0.0039 %/Mercury 0.0082 %/Indium 0.0098 %/Bismuth 0.0101 %/Cadmium 0.0124 %/Arsenic 0.017 %/Silver 0.0201 %/Antimony 0.0245 %/Copper 0.0245 %/Tin 60.02 %/Gold (0.0004) %/Zinc (0.0004) %/Tellurium (0.0005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction