Learn More
Syngene™ LED Module for Syngene Gbox Chemi System
Syngene™ LED Module for Syngene Gbox Chemi System for western fluorescence
Brand:  Syngene™ LED-BLUE-M x 2
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15819962
  • 1305.00€ / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Specifications
Blue
Gbox Chemi Systems
LED Module, Multiplex
2
465nm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products