Learn More
BINDER™ Access Port with Silicone Plug, Left Access
Product Code. 15878806 Shop All BINDER Products
Click to view available options
Material:
Steel
Unit Size:
Each
Product Code. 15878806
Brand: BINDER™ 80121334
Only null left
Add to basket
Add to basket
BINDER™ Left Access Port is an accessory for use on the left of a variety of chambers and incubators.
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 80 mm, left
For Use With (Equipment) BINDER Chambers Series MK
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 80 mm