Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: KBF 240, KBF-S 240, KBF-720, KBF-S 720, KBF 1020, KBF 115
Brand:  BINDER™ 8012-1442
Additional Details : Weight : 0.31000kg
Product Code. 16444229
  • 505.00€ / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 30 mm, left
Access Port
30 mm
BINDER Chambers Series KBF
Silicone plug