Learn More
KD Scientific Legato™ 950 OEM Syringe Pump
Product Code. 16343747
Click to view available options
Number of Channels:
1
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 16343747
Brand: KD Scientific 788950
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
The Legato™ 950 is ideal for applications interfacing to a computer. Accommodates syringes 0.5 μL to 60 mL. User definable flow rates with selectable target volume or time values to control the total infusion volume. Has the ability to interface and integrate into other systems.
  • ± 0.5% Accuracy
  • 0.5 μL to 60 mL syringes
  • Continuous mode of operation
  • Programmable with ASCII commands
  • Encoder stall detection
  • USB Interface
  • TTL Interface
  • Customizable syringe mechanisms available
  • Customizable chassis designs available
  • Advanced micostepping techniques
  • Linear force 30 lb. (13.6 kg)
  • CE, CB Scheme, EU RoHS compliant
Specifications
Voltage 100/240 V
Flow Rate 1.28 pL/min. to 88.28 mL/min.
Certifications/Compliance CE, UL, CSA, CB Scheme, EU RoHS
Linear Force (Metric) 13.6 kg (30 lb) @ 100% Force Selection
Temperature (Metric) Storage -10°C to +70°C
Type Syringe Pump
Display Type Touchscreen
Flow Rate Accuracy ±0.5%
Number of Channels 1
Temperature (Metric) Max 40
Temperature (Metric) Min. 4