missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Leucomalachite Green
Click to view available options
Quantity:
2 g
10 g
Descrizione
Leucomalachite Green is a reagent for the quantitative colorimetric micro determination of hemoglobin and other heme compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Melting Point | 100°C to 102°C |
Beilstein | 2140506 |
Merck Index | 14,5699 |
Solubility Information | Soluble in water (<0.1mg/ml),chloroform,benzene,ethyl ether,ethanol and toluene. |
Formula Weight | 330.47 |
Sensitivity | Air Sensitive |
Quantity | 2 g |
Physical Form | Powder |
Chemical Name or Material | Leucomalachite Green |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'