Learn More
Greiner Bio-One Leucosep Polypropylene Round Bottom Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
Brand:  Greiner Bio-One 227288
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10030322
  • 4885.00€ / Pack of 250
Estimated Shipment: 16-04-2024
to see stock.
Add to basket
Description
Description
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes
  • No recontamination with erythrocytes
  • Service Plan
    No
Specifications
Specifications
50 mL
Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow
Aseptically Manufactured
Centrifuge Tube
Polypropylene
Porous barrier
Round