Learn More
Greiner Bio-One Leucosep Polypropylene Centrifugation Tubes
Click to view available options
Capacity (Metric):
12 mL
50 mL
Sterility:
Aseptically Manufactured
Sterile
Bottom Shape:
Conical
Round
Unit Size:
Pack of 250
Pack of 500
Description
- Enrichment directly from whole blood
- Simplified filling through porous barrier
- Rapid separation in 15 minutes at room temperature
- Removal of erythrocytes and granulocytes
- No recontamination with erythrocytes
-
Service Plan
No
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Polypropylene |
| For Use With | Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow |
| Includes | Porous barrier |
| Sterility | Aseptically Manufactured |
| Bottom Shape | Conical |
| Product Type | Centrifuge Tube |