Learn More
Elma™ Level Sets
Product Code. p-200004986
Click to view available options
Model:
L
M
S
XL
Unit Size:
1 set
Each
Product Code. 17891327
Brand: Elma™ 1125225
Only null left
Add to basket
Add to basket
Modular basket system for the Elmasonic P, Elmasonic Select, and EASY.
  • Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Type Level Set
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Model S