Learn More
Nickel Electro™ Lid for 15L Stainless Steel Bucket
For use with 15L stainlees steel buckets
Brand: Nickel Electro™ BUCS-L150
23.44 EUR valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- High quality stainless steel construction
Specifications
Lid | |
Yes | |
Lid | |
Bright Polished |
Stainless Steel | |
15 L Stainless Steel Bucket | |
Ea. |