Learn More
Cytiva Lid Kit for Funnel
Product Code. 17174281
Change view
Click to view available options
:
For 300 mL Funnel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17174281 For 300 mL Funnel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17174281 Supplier Cytiva Supplier No. 4246
Only null left
Add to basket
Add to basket
Replacement lid kit for funnels with unique magnetic seal.
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 300 mL Funnel
Material PES, Polypropylene
Sterility Non-sterile
Includes Silicone O-Ring