missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine EP Impurity D (as Hydrochloride), Mikromol™

Product Code. 17981606
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17981606

Brand: Mikromol MM0102.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lidocaine EP Impurity D (as Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Norlidocaine Hydrochloride (Monoethylglycinexylidide Hydrochloride)
CAS 7729-94-4
Analyte or Component Names Norlidocaine Hydrochloride (Monoethylglycinexylidide Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C12 H18 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym N-(2,6-Dimethylphenyl)-2-(ethylamino)acetamide Hydrochloride (Monoethylglycinexylidide Hydrochloride), Lidocaine Hydrochloride Imp. D (EP), Lidocaine Imp. D (EP), N-(2,6-Dimethylphenyl)-2-(ethylamino)acetamide Hydrochloride, Lidocaine Hydrochloride Imp. D (EP) as Hydrochloride, Monoethylglycinexylidide Hydrochloride, Norlidocaine Hydrochloride, Lidocaine Hydrochloride Impurity D as Hydrochloride, Lidocaine Impurity D as Hydrochloride
InChI Formula InChI=1S/C12H18N2O.ClH/c1-4-13-8-11(15)14-12-9(2)6-5-7-10(12)3;/h5-7,13H,4,8H2,1-3H3,(H,14,15);1H
SMILES Cl.CCNCC(=O)Nc1c(C)cccc1C
IUPAC Name N-(2,6-dimethylphenyl)-2-(ethylamino)acetamide;hydrochloride
Molecular Weight (g/mol) 242.75
Quantity 100 mg
Formula Weight 242.1186 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.