Learn More
BOLA Lids
Click to view available options
Diameter (Metric) Outer:
60 mm
66 mm
82 mm
125 mm
166 mm
185 mm
Unit Size:
Each
Description
Lid with centering shoulder.
Specifications
Specifications
| Diameter (Metric) Outer | 66 mm |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Certifications/Compliance | FDA |