Learn More
Abeba™ Light 1027 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1027/39
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Microfiber black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Microfiber | |
Slip-resistant | |
1027 |
Black | |
39 | |
Polyurethane | |
Steel |