Learn More
Abeba™ Light 1032 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1032/46
Additional Details : Weight : 1.12500kg
 View more versions of this product
Product Code. 15626737
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Leather
Slip-resistant
1032
White
46
Polyurethane
Steel