Learn More
Abeba™ Light 1035 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1035/42
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20345:2011 SB A, E, FO SRA
- Smooth leather
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3556)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRA | |
Leather | |
Slip-resistant | |
1035 |
Black | |
42 | |
Polyurethane | |
Steel |