Learn More
Abeba™ Light 1035 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1035/46
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15666747
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRA
  • Smooth leather
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3556)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area with elastic band
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRA
Leather
Slip-resistant
1035
Black
46
Polyurethane
Steel