Learn More
Abeba™ Light 1127 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1127/38
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 15666997
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRA
  • Microfiber black
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRA
Microfiber
Slip-resistant
1127
Black
38
Polyurethane