Learn More
Abeba™ Light 1136 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand:  Abeba™ 1136/41
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15607067
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Double velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRA
Leather
Slip-resistant
1136
Black
41
Polyurethane