Learn More
Abeba™ Light 1136 Shoes
Product Code. 15647057
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15647057
Brand: Abeba™ 1136/35
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
  • CE EN ISO 20347:2012 O1 FO SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • Antistatic
  • Breathable lining with Silver ions
  • Double velcro fastening
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O1 FO SRA
Color Black
Material Leather
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 1136