Learn More
Abeba™ Light 1136 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1136/36
Additional Details : Weight : 0.80000kg
Description
- CE EN ISO 20347:2012 O1 FO SRA
- Leather with Air-Mesh textile inlays
- Black
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- Antistatic
- Breathable lining with Silver ions
- Double velcro fastening
Specifications
CE EN ISO 20347:2012 O1 FO SRA | |
Leather | |
Slip-resistant | |
1136 |
Black | |
36 | |
Polyurethane |