Learn More
Abeba™ Light 1160 Shoes
Special lining with silver ions absorbs resperation. Includes lightweight PU-sole with high slip resistance.
Brand: Abeba™ 1160/42
Additional Details : Weight : 0.60000kg
Description
- CE EN ISO 20347:2012 OB A, E, FO SRA
- Smooth leather
- White/Red
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3558)
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
Specifications
CE EN ISO 20347:2012 OB A, E, FO SRA | |
Leather | |
Slip-resistant | |
1160 |
White, Red | |
42 | |
Polyurethane |