Learn More
Abeba™ Light 31026 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 31026/44
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Microfiber
- White
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Microfiber | |
Slip-resistant | |
31026 |
White | |
44 | |
Polyurethane | |
Steel |