Learn More
Abeba™ Light 31047 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 31047/43
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20345:2011 S2 SRA
- Smooth leather white
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3557)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver Point
- Adjustable instep area
- Double velcro fastening
- Overcap with scratch-resistant coating
- Suitable for heel and foot lift
Specifications
CE EN ISO 20345:2011 S2 SRA | |
Leather | |
Slip-resistant | |
31047 |
White | |
43 | |
Polyurethane | |
Steel |