Learn More
Abeba™ Light 31047 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 31047/46
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15638417
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20345:2011 S2 SRA
  • Smooth leather white
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver Point
  • Adjustable instep area
  • Double velcro fastening
  • Overcap with scratch-resistant coating
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRA
Leather
Slip-resistant
31047
White
46
Polyurethane
Steel