Learn More
Abeba™ Light 31056 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
69.65€
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S1 SRA
Color Black
Material Leather
Sole Features Slip-resistant
Sole Material Polyurethane
View More Specs
Products 14
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15648447
View Documents Promotion Details
Abeba™
31056/35
35 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15658447
View Documents Promotion Details
Abeba™
31056/36
36 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15668447
View Documents Promotion Details
Abeba™
31056/37
37 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15678447
View Documents Promotion Details
Abeba™
31056/38
38 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15688447
View Documents Promotion Details
Abeba™
31056/39
39 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15698447
View Documents Promotion Details
Abeba™
31056/40
40 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15608457
View Documents Promotion Details
Abeba™
31056/41
41 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15618457
View Documents Promotion Details
Abeba™
31056/42
42 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15628457
View Documents Promotion Details
Abeba™
31056/43
43 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15638457
View Documents Promotion Details
Abeba™
31056/44
44 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15648457
View Documents Promotion Details
Abeba™
31056/45
45 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15658457
View Documents Promotion Details
Abeba™
31056/46
46 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15668457
View Documents Promotion Details
Abeba™
31056/47
47 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15678457
View Documents Promotion Details
Abeba™
31056/48
48 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description
Description
  • CE EN ISO 20345:2011 S1 SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3557)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver Point
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRA
Leather
Polyurethane
Steel
Black
Slip-resistant
31056