Learn More
Abeba™ Light 31127 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 31127/37
Additional Details : Poids : 0.90000kg
Description
- CE EN ISO 20347:2012 O2 FO SRA
- Microfiber black
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
Specifications
CE EN ISO 20347:2012 O2 FO SRA | |
Microfiber | |
Slip-resistant | |
31127 |
Black | |
37 | |
Polyurethane |