Learn More
Abeba™ Light 31136 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 31136/42
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15668667
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRA
  • Leather with Air-Mesh textile inlays
  • Black
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Double velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRA
Leather
Slip-resistant
31136
Black
42
Polyurethane