Learn More
Abeba™ Light 31151 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand: Abeba™ 31151/40
Additional Details : Weight : 0.95900kg
Description
- CE EN ISO 20347:2012 O2 FO SRA
- Smooth leather white
- Slip resistance PU outsole sole
- Replaceable acc Wave insole (article no. 3559)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Temperature regulating lining Comfortemp™ with mPCM-Technology
- adjustable instep area
Specifications
CE EN ISO 20347:2012 O2 FO SRA | |
Leather | |
Slip-resistant | |
31151 |
White | |
40 | |
Polyurethane |