Learn More
Abeba™ Light 31151 Shoes
ESD dissipative shoe includes special respiration absorbing lining for the foot to breathe.
Brand:  Abeba™ 31151/40
Additional Details : Weight : 0.95900kg
 View more versions of this product
Product Code. 15658697
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O2 FO SRA
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3559)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Temperature regulating lining Comfortemp™ with mPCM-Technology
  • adjustable instep area
Specifications
Specifications
CE EN ISO 20347:2012 O2 FO SRA
Leather
Slip-resistant
31151
White
40
Polyurethane