Learn More
Invitrogen™ EVOS™ Light Cube, AO Red
Product Code. 11927305
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 11927305
Brand: Invitrogen™ AMEP4671
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
  • Contains unique, proprietary LED light cubes
  • Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
  • Each cube contains an LED, condensing optics, and hard-coated filters
  • User-changeable and automatically recognized by system
  • Shorter light path that affords superior detection of fluorescent signals
  • Continuous illumination that enables consistent results
  • 50,000+ hour bulb lifetime that affords lower operating costs
  • Adjustable light intensity that helps reduce photobleaching
TRUSTED_SUSTAINABILITY
Specifications
Type Light Cube
Product Line EVOS
For Use With (Equipment) Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-Coated Filters, LED
Wavelength Ex: 442/46 Em: 628/32
Light Source Metal Halide (Bulb)
Quantity 1 Ea.