Learn More
Tork Light Duty Cleaning Cloth Combi-Roll W1/W2/W3
Shop All Tork Products
Click to view available options
Color:
White
Unit Size:
1 roll
Description
- Multipurpose cleaning cloths for light-duty cleaning tasks.
- Use these durable cloths wet or dry to clean dust and dirt, and you can rinse and reuse them as needed.
- These disposable cleaning cloths are compatible with most chemical solvents, so no need to change your cleaning products.
- This combi-roll has 600 sheets to avoid runouts and save refill time.
Specifications
Specifications
| Product Type | Cleaning Cloth |
| Color | White |
| Ply | 1-ply |