missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Description
Alkaline phosphatase (AP) is a hydrolyase enzyme that is frequently conjugated to antibodies for use in immunoassays. PNPP is a popular colorimetric substrate for AP; the product of the reaction, p-nitrophenol, can be read at 405nM. Fluorogenic substrates (e.g. 4-methylumbelliferyl phosphate; MUP) may also be employed.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 1 mg |
| Conjugate | Alkaline Phosphatase |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
| Product Type | Alkaline Phosphatase Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction