missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid DyLight 633 Antibody Labeling Kit

Product Code. 18028465 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1mg
300µg
30µg
This item is not returnable. View return policy

Product Code. 18028465

Brand: Novus Biologicals™ 3250030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

DyLight 594 generates red fluorescence for a wide array of fluorescence labeling-based applications. It has a strong absorption at 593nm, high fluorescence at 618nm (extinction coefficient 8.0 x10 cm-1M-1) and high quantum yield.
  • Lightning-Link™ Rapid technology can be used to label small quantities of antibody with 100% recovery
  • Easy to use antibody labeling kit with quick labeling times
  • From 10μg up to 2mg
  • Kit with 30 seconds hands-on time
  • No separation steps and no losses
  • Scalable technology
Applications:
  • Western Blotting, ELISA, Immunohistochemistry, Immunofluorescence and FACS analysis

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate DyLight 633
Product Type Rapid DyLight 633 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.