missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liothyronine Methyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Liothyronine Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Liothyronine Methyl Ester |
| CAS | 54914-53-3 |
| Analyte or Component Names | Liothyronine Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 I3 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Liothyronine Methyl Ester, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-L-Tyrosine methyl ester, 3,5,3'-Triiodo-L-thyronine methyl ester, L-Triiodothyronine methyl ester |
| InChI Formula | InChI=1S/C16H14I3NO4/c1-23-16(22)13(20)6-8-4-11(18)15(12(19)5-8)24-9-2-3-14(21)10(17)7-9/h2-5,7,13,21H,6,20H2,1H3/t13-/m0/s1 |
| SMILES | COC(=O)[C@@H](N)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1 |
| IUPAC Name | methyl (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction