Learn More
Sampling Systems™ Liquid Dip Sampler
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
Unit Size:
Each
Description
- Made from high quality and chemically resistant 316L stainless steel
- Different sizes available
- Convenient, hands-free emptying feature
Specifications
Specifications
| Product Type | Dip Sampler |
| Material | Stainless Steel (316L) |
| Autoclavable | Autoclavable |
| Capacity (English) | 3.4 oz. |
| Capacity (Metric) | 100 mL |
| Disposable | Reusable |
| Graduated | No |
| Sterility | Non-sterile |