missing translation for 'onlineSavingsMsg'
Learn More

CD31/PECAM-1 Antibody (C31.10), Biotin , R&D Systems™

Code produit 18611297 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18611297 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18611297 Fournisseur Novus Biologicals Code fournisseur NBP311580B
 Article restreint
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigène CD31/PECAM-1
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C31.10
Conjugué Biotin
Formule PBS
Alias de gène adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Espèces hôtes Mouse
Immunogène Recombinant human CD31/PECAM-1 protein (Uniprot: P16284)
Méthode de purification Protein A or G purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 5175
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Type de produit Antibody
Forme Purified
Isotype IgG1 κ
Gamme de produits Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.