missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, TraceSELECT™, ≥99.998% (metals basis)

Product Code. p-7120519 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Poly Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15686040

Brand: Honeywell-Fluka 6246225G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Color White
pH 9.0 to 10.00 (at 1g/L, 20°C)
Physical Form Crystalline
Quantity 25 g
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
IUPAC Name dilithium;carbonate
Formula Weight 73.89g/mol
Percent Purity ≥99.998% (metals basis)
Grade TraceSELECT™
Odor Odorless
Packaging Poly Bottle
Density 2.11g/cm3
Chemical Name or Material Lithium carbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.