missing translation for 'onlineSavingsMsg'
Learn More

Lithium fluoride, 97%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4510020
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]
This item is not returnable. View return policy
7789-24-4
FLi
25.94
MFCD00011090
PQXKHYXIUOZZFA-UHFFFAOYSA-M
lithium fluoride, fluorolithium, lithium monofluoride, trilithium trifluoride, lithium fluoride li3f3, lithium fluorure, ntl 50, lithium fluoride 6lif, lithium fluoride 7lif
224478
[Li+].[F-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1681.0°C
Melting Point 845.0°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min
Linear Formula LiF
Fieser 01,933
Merck Index 15,5587
Solubility Information Solubility in water: 0.29 g/100mL (20°C). Other solubilities: soluble in acids, insoluble in alcohol
IUPAC Name lithium;fluoride
Formula Weight 25.93
Percent Purity 97%
Grade Analytical
Loss on Drying 0.4% (200°C) (typical)
Packaging Plastic bottle
Specific Gravity 2.63
Density 2.6300g/mL
Chemical Name or Material Lithium fluoride
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'