missing translation for 'onlineSavingsMsg'
Learn More

Lithium hexafluoroarsenate(V), 99%, Thermo Scientific Chemicals

Catalog Number p-7048807
92.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
29935-35-1
AsF6Li
195.85
MFCD00011076
YYPUQABLGAAHDV-UHFFFAOYSA-H
lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
9837036
[Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
29935-35-1
AsF6Li
195.85
MFCD00011076
YYPUQABLGAAHDV-UHFFFAOYSA-H
lithium hexafluoroarsenate, lithium hexafluoroarsenate v, lithium hexafluoroarsoranuide, lithium hexakis fluoranyl arsenic 1-, lithotab hexafluoro-$l^ 5-arsanuide, lithium 1+ hexafluoro-??-arsanuide
9837036
[Li+].[F-].[F-].[F-].[F-].[F-].[F-].[As+5]

Lithium hexafluoroarsenate(V) is a potential battery electrolyte material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
UN Number UN1557
Solubility Information Insoluble in water.
IUPAC Name lithium;hexafluoroarsenic(1-)
Formula Weight 195.85
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material Lithium hexafluoroarsenate(V)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'