missing translation for 'onlineSavingsMsg'
Learn More

Lithium nitrate, anhydrous, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048595
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
This item is not returnable. View return policy
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
lithium;nitrate
[Li+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7790-69-4
LiNO3
68.94
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
10129889
CHEBI:63315
lithium;nitrate
[Li+].[O-][N+]([O-])=O

Lithium nitrate, anhydrous is used in the manufacture of red-colored fireworks and flares. has been proposed as a medium to store heat collected from the sun for cooking. A Fresnel lens would be used to melt solid lithium nitrate, which would then function as a 'solar battery', allowing heat to be redistributed later by convection

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 600°C (decomposition)
Melting Point 264°C
Physical Form Crystalline
Quantity 1 g
UN Number UN2722
Merck Index 14,5536
Solubility Information Soluble in water. Also soluble in ethanol,methanol,pyridine,ammonia and acetone.
Formula Weight 68.95
Percent Purity 99.98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.38 g/mL
Assay (metals basis)
Chemical Name or Material Lithium nitrate, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'