missing translation for 'onlineSavingsMsg'
Learn More

Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049129
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
10377-48-7
Li2O4S
109.94
MFCD00011086
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
66320
CHEBI:53474
[Li+].[Li+].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10377-48-7
Li2O4S
109.94
MFCD00011086
INHCSSUBVCNVSK-UHFFFAOYSA-L
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
66320
CHEBI:53474
[Li+].[Li+].[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 130°C (in -H2O)
Color White
Physical Form Crystalline
Quantity 5 g
Linear Formula Li2SO4·xH2O
Merck Index 14,5541
IUPAC Name dilithium;sulfate
Formula Weight 109.94 (Anhydrous)
Percent Purity 99.996%
Grade Puratronic™
Density 2.06 g/mL
Assay (metals basis)
Chemical Name or Material Lithium sulfate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'